Order Entry
Ireland
ContactUsLinkComponent
Anti-VEGF Rabbit Recombinant Antibody [clone: BEVACIZUMAB]
Anti-VEGF Rabbit Recombinant Antibody [clone: BEVACIZUMAB]
  ABNORAB01192.200UG
 :  Abnova
Anti-VEGF Rabbit Recombinant Antibody [clone: BEVACIZUMAB]
  ABNORAB01192.200UG
 :  Abnova

 

  • Antigen symbol:
    VEGF
  • Clonality:
    Recombinant
  • Clone:
    BEVACIZUMAB
  • Host:
    Rabbit
  • Reactivity:
    Human
  • Immunogen:
    Recombinant rabbit monoclonal antibody raised against human VEGF.
  • Pk:
    200 µG