Anti-IgE Rabbit Recombinant Antibody [clone: TES-C21]
Catalog # ABNORAB01168.200UG
Supplier: Abnova
Specifications
- Antigen symbol:IgE
- Clonality:Recombinant
- Clone:TES-C21
- Host:Rabbit
- Reactivity:Human
- Immunogen:Recombinant rabbit monoclonal antibody raised against human IgE.
- Pk:200 µG