Anti-Integrin alpha 4 Mouse Recombinant Antibody [clone: NATALIZUMAB]
Catalog # ABNORAB01173.200UG
Supplier: Abnova
Specifications
- Antigen symbol:Integrin alpha 4
- Clonality:Recombinant
- Clone:NATALIZUMAB
- Host:Mouse
- Reactivity:Human
- Immunogen:Recombinant mouse monoclonal antibody raised against human Integrin alpha 4.
- Pk:200 µG