Anti-RSV Human Recombinant Antibody [clone: RSHZ19 (FELVIZUMAB)]
Catalog # ABNORAB01177.200UG
Supplier: Abnova
Specifications
- Antigen symbol:RSV
- Clonality:Recombinant
- Clone:RSHZ19 (FELVIZUMAB)
- Host:Human
- Immunogen:Recombinant human monoclonal antibody raised against RSV.
- Pk:200 µG