Anti-TNF alpha Mouse Recombinant Antibody [clone: CA2 (INFLIXIMAB)]
ABNORAB01182.200UG
: Abnova
- Antigen symbol:TNF alpha
- Clonality:Recombinant
- Clone:CA2 (INFLIXIMAB)
- Host:Mouse
- Reactivity:Human
- Immunogen:Recombinant mouse monoclonal antibody raised against human TNF alpha.
- Pk:200 µG