Order Entry
Ireland
ContactUsLinkComponent
Anti-CD4 Mouse Recombinant Antibody [clone: CE9.1 (CLENOLIXIMAB)]
Anti-CD4 Mouse Recombinant Antibody [clone: CE9.1 (CLENOLIXIMAB)]
Catalog # ABNORAB01143.200UG
Supplier:  Abnova
Anti-CD4 Mouse Recombinant Antibody [clone: CE9.1 (CLENOLIXIMAB)]
Catalog # ABNORAB01143.200UG
Supplier:  Abnova

Specifications

  • Antigen symbol:
    CD4
  • Clonality:
    Recombinant
  • Clone:
    CE9.1 (CLENOLIXIMAB)
  • Host:
    Mouse
  • Reactivity:
    Human
  • Immunogen:
    Recombinant mouse monoclonal antibody raised against human CD4.
  • Pk:
    200 µG

Specifications

About this item