Anti-CD40L Mouse Recombinant Antibody [clone: HU5C8 (RUPLIZUMAB)]
ABNORAB01144.200UG
: Abnova
- Antigen symbol:CD40L
- Clonality:Recombinant
- Clone:HU5C8 (RUPLIZUMAB)
- Host:Mouse
- Reactivity:Human
- Immunogen:Recombinant mouse monoclonal antibody raised against human CD40L.
- Pk:200 µG