Anti-CDK4 Mouse Monoclonal Antibody [clone: P6D10AT]
Catalog # ABNOMAB23652.100UG
Supplier: Abnova
Specifications
- Antigen symbol:CDK4
- Clonality:Monoclonal
- Clone:P6D10AT
- Host:Mouse
- Reactivity:Human
- Immunogen:Mouse monoclonal antibody raised against synthetic peptide of human CDK4, clone P6D10AT.
- Pk:100 µG