Anti-VCAN DNA Rabbit Polyclonal Antibody
ABNOH00001462-W01P
: Abnova
- Antigen symbol:VCAN DNA
- Clonality:Polyclonal
- Host:Rabbit
- Reactivity:Human
- Immunogen:Rabbit polyclonal antibody raised against a partial-length human VCAN DNA using DNAx™ Immune technology
- Pk:100 µG