Anti-PLK1 (pThr210) Rabbit Monoclonal Antibody [clone: C2] (FITC)
ABNOMAB23499.100
: Abnova
- Antigen symbol:PLK1
- Clonality:Monoclonal
- Clone:C2
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Host:Rabbit
- Reactivity:Human
- Modification:pThr210
- Immunogen:Rabbit monoclonal antibody raised against synthetic phosphopeptide of human PLK1.
- Pk:100 AU