Order Entry
Ireland
ContactUsLinkComponent
CSR Infinity H₂ Purifier
CSR Infinity H₂ Purifier
Catalog # SGEA202381-SS
CSR Infinity H₂ Purifier
Catalog # SGEA202381-SS
Specifications
  • Description:
    Infinity H₂ purifier, ¹/₄" stainless steel compression fittings
  • Pk:
    1
Specifications
About this item
The CRS Infinity H2 purifier is a self regenerating purifier that removes oxygen from hydrogen at ambient temperature.
  • High quality 316 stainless steel construction
  • For use at ambient temperatures
  • For use with hydrogen only
The reaction consumes the oxygen and creates an excess of water vapor. When paired with an additional moisture filter it is the perfect solution for hydrogen generators. This catalyic purifier has the potential to last forever, however the media can be easily contaminated by oil, organic solvents, heavy metals, sulfur compounds etc.