Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Ireland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Seals
Gaskets
Gasket for Flat Door
Gasket for Flat Door
Gasket for Flat Door
Catalog # TUTTGAS080-0310
Supplier:
TUTTNAUER
Expand image
Gasket for Flat Door
Catalog # TUTTGAS080-0310
Supplier:
TUTTNAUER
Specifications
Ø:
229 mm
Description:
Gasket for flat door
Material:
Silicone
Pk:
1
Specifications
About this item
Product Details
Product Details