Indoor Outdoor Vinyl Labels for M410, BMP41, M510, BMP51, BMP53 and M511 Printers
: Brady
Make labels that withstand up to 10 years of outdoor abuse with this low-shrink vinyl label material (B-595). Print almost anywhere with a durable Brady handheld label printer.
- Outdoor durable
- Low temperature resistant
- Fuel and oil resistant
- High adhesion
- Abrasion resistant
- Gloss finish
Ideal for labelling pipes, equipment, shelves and other industrial surfaces. Designed to withstand prolonged exposure to UV light, chemicals, greases and oils, salt spray, cleaners and detergents, and grime. Made with high-tack, permanent acrylic adhesive for use on rough, highly-textured, dirty and smooth surfaces alike, especially in cold temperatures.
Gloss finish, abrasion-resistant, chemical-resistant, oil-resistant, water-resistant B-595 labels are compatible with Brady printer models M410, BMP41, M510, BMP51, BMP53, and M511.
: RoHS compliant to 2011/65/EU (Pass 2015/863).
Frequently Bought Together