Order Entry
Ireland
ContactUsLinkComponent
Milli-Q® HX 7040/7080/7120/7150 SD Water Purification Systems
Milli-Q® HX 7040/7080/7120/7150 SD Water Purification Systems
Supplier:  Merck
Certificates
undefined
Milli-Q® HX 7040/7080/7120/7150 SD Water Purification Systems
Supplier:  Merck

Specifications

  • Catalog No:
  • MILLZLXLSD52040
  • MILLZLXLSD52080
  • MILLZLXLSD52150
  • MILLPR0GTXLCS1
  • MILLZLXLSD52120
  • MILLPR0GTXLCS2
  • Model:
  • Milli-Q® HX 7040 SD
  • Milli-Q® HX 7080 SD
  • Milli-Q® HX 7150 SD
  • Milli-Q® HX 7120 SD
  • Feed water:
  • Potable tap water (consult manual for requirements)
  • Potable tap water (consult manual for requirements)
  • Potable tap water (consult manual for requirements)
  • Potable tap water (consult manual for requirements)
  • Flow rate:
  • 40 L/h
  • 80 L/h
  • 150 L/h
  • 120 L/h
  • Conductivity:
  • <0,2 μS/cm (at 25 °C)
  • <0,2 μS/cm (at 25 °C)
  • <0,2 μS/cm (at 25 °C)
  • <0,2 μS/cm (at 25 °C)
  • TOC content:
  • <30 ppb
  • <30 ppb
  • <30 ppb
  • <30 ppb
  • Resistivity at +25 °C:
  • >5 MΩ.cm
  • >5 MΩ.cm
  • >5 MΩ.cm
  • >5 MΩ.cm
  • Bacteria:
  • <10 CFU/ml
  • <10 CFU/ml
  • <10 CFU/ml
  • <10 CFU/ml
  • Weight:
  • 97 kg, dry; 116 kg, shipping
  • 106 kg, dry; 125 kg, shipping
  • 126 kg, dry; 145 kg, shipping
  • 114 kg, dry; 133 kg, shipping

Specifications

Frequently Bought Together