Sentinel XT® Clear Full Hood
Catalog # ILCDS-3112-30
Supplier: ILC DOVER
Specifications
- Description:Sentinel XT® clear full hood
- Size:Universal
- Pk:30
Specifications
About this item
The Sentinel XT® Clear Full Hood is a superior solution for protection in the pharmaceutical industry when compared to other PAPR masks and facepieces.
- For use with the Sentinel XT® Blower system
- 320 degree field of view
- No fit test required
- Save time and storage space with just one size needed
- APF of 1000
- Latex-free
With an APF of 1000, the clear full hood offers a 320 degree field of view, allowing the wearer to have facial hair and/or a head covering, and reducing feelings of claustrophobia. No headband or ratchet harness is needed to secure the hood, ensuring a more comfortable experience. Metal clips allow the outer bib to secure to protective clothing.