

2-Propanol SLSI, Electronic Grade
Catalog # 50489128.320
Supplier: VWR Chemicals
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:2,5 L
- Seq:5096
Specifications
Specification Test Results
- Assay (GC):≥ 99.90 %
- Identification by GC :Conforms
- Particle size (>0.50 μm)(/ml)(at filling):≤ 100
- Resistivity (MOhm-cm):≥ 20
- Water:≤ 200 ppm
- Cl (Chloride):≤ 100 ppb
- PO₄ (Phosphate):≤ 100 ppb
- SO₄ (Sulphate):≤ 100 ppb
- Ag (Silver):≤ 1.00 ppb
- Al (Aluminium):≤ 1.00 ppb
- As (Arsenic):≤ 1.00 ppb
- Au (Gold):≤ 1.00 ppb
- B (Boron):≤ 1.00 ppb
- Ba (Barium):≤ 1.00 ppb
- Be (Beryllium):≤ 1.00 ppb
- Bi (Bismuth):≤ 1.00 ppb
- Ca (Calcium):≤ 1.00 ppb
- Cd (Cadmium):≤ 1.00 ppb
- Co (Cobalt):≤ 1.00 ppb
- Cr (Chromium):≤ 1.00 ppb
- Cu (Copper):≤ 1.00 ppb
- Fe (Iron):≤ 1.00 ppb
- Ga (Gallium):≤ 1.00 ppb
- Ge (Germanium):≤ 1.00 ppb
- In (Indium):≤ 1.00 ppb
- K (Potassium):≤ 1.00 ppb
- Li (Lithium):≤ 1.00 ppb
- Mg (Magnesium):≤ 1.00 ppb
- Mn (Manganese):≤ 1.00 ppb
- Mo (Molybdenum):≤ 1.00 ppb
- Na (Sodium):≤ 2.00 ppb
- Ni (Nickel):≤ 1.00 ppb
- Pb (Lead):≤ 1.00 ppb
- Pt (Platinum):≤ 1.00 ppb
- Sb (Antimony):≤ 1.00 ppb
- Sn (Tin):≤ 1.00 ppb
- Sr (Strontium):≤ 1.00 ppb
- Ti (Titanium):≤ 1.00 ppb
- Tl (Thallium):≤ 1.00 ppb
- V (Vanadium):≤ 1.00 ppb
- Zn (Zinc):≤ 1.00 ppb
- Zr (Zirconium):≤ 1.00 ppb
Specification Test Results
About this item
:
:60.1 g/mol
Boiling Pt:82 °C (1013 hPa)
Melting Pt:–89 °C
Density:0,786 g/cm³ (20 °C)
Flash Pt:12 °C
Storage Temperature:Ambient
EINECS:200-661-7
UN:1219
ADR:3,II
REACH:01-2119457558-25
:GHS02
:GHS07
CAS Number:67-63-0
Formula:(CH₃)₂CHOH
MDL Number:MFCD00011674