Order Entry
Ireland
ContactUsLinkComponent
Manual Vial Crimper
Manual Vial Crimper
Catalog # ADLM6030555-1
undefined
Manual Vial Crimper
Catalog # ADLM6030555-1

Specifications

  • Description:
    Manual vial crimper
  • W×D×H:
    305×254×457 mm
  • Pk:
    1

Specifications

About this item

A durable and efficient, manually operated, benchtop vial crimping machine.

  • Designed for 'plain', 'flip top/flip off', and 'flip tear up' seals
  • Crimping and de-capping of 13, 18 and 20 mm seals
  • Fast and reliable, for a professional quality finish
  • Easy to set-up and use, to get started straight away
  • Corrosion-resistant construction for long service life
  • Weight: 9,5 kg

Heads are quickly and easily changed without the use of tools, since they simply screw in and out. The unit is height and position-adjustable, for operator comfort and safety.

All crimping and de-capping heads are interchangeable with the Adelphi Manufacturing powered vial crimper.

Delivery: Supplied without head (available to purchase separately), with stainless steel bench stand.