Order Entry
Ireland
ContactUsLinkComponent
Greenough Stereo Microscopes, Ivesta 3
Greenough Stereo Microscopes, Ivesta 3
Supplier:  LEICA MICROSYSTEMS
Certificates
undefined
Greenough Stereo Microscopes, Ivesta 3
Supplier:  LEICA MICROSYSTEMS
Specifications
  • Catalog No:
  • 630-3533
  • 630-3527
  • 630-3536
  • 630-3531
  • 630-3535
  • 630-3534
  • 630-3538
  • 630-3530
  • 630-3537
  • Model:
  • Ivesta 3 (Integrated camera)
  • Ivesta 3 (C-mount)
  • Ivesta 3
  • Objectives:
  • Apochromatic 0,5×, 0,63×, 0,75×, 1,6×, 2,0× (lead-free)
  • Apochromatic 0,5×, 0,63×, 0,75×, 1,6×, 2,0× (lead-free)
  • Apochromatic 0,5×, 0,63×, 0,75×, 1,6×, 2,0× (lead-free)
  • Magnification:
  • 6,1× - 55×
  • 6,1× - 55×
  • 6,1× - 55×
  • Zoom lens:
  • 9:01
  • 9:01
  • 9:01
  • Working distance:
  • 122 mm
  • 122 mm
  • 122 mm
  • Camera:
  • Live image on an HDMI monitor at up to 60 fps (3840×2160 pixels);
    Full-screen image capture at 12 MP;
    File formats JPG, TIF, BMP, MJPG; for overlays also PNG
  • Sensor:
  • 7,81 mm (type 1/2,3" CMOS), diagonal
  • Resolution:
  • 500 lp/mm (ma×.)
  • 500 lp/mm (ma×.)
  • 500 lp/mm (ma×.)
  • Output:
  • 4× USB 2 connectors, type A; HDMI 2.0a, HDMI plug type A
  • 50% video/50% visual, permanent (video/photo outlet)
  • Software:
  • Enersight (supported)
  • Weight:
  • 2,2 kg
  • 1,8 kg
  • 1,6 kg
  • Optical design:
  • 10° Greenough using best-corrected central part of the objective (lead-free)
  • 10° Greenough using best-corrected central part of the objective (lead-free)
  • 10° Greenough using best-corrected central part of the objective (lead-free)
Specifications
Related Information
Components
Product Information
Frequently Bought Together