Cole-Parmer® WB-300 Series General-Purpose Digital Water Baths, Stuart
Supplier: Cole Parmer
Certificates
About this item
Easy to use and easy to clean.
- Analogue dial with digital temperature display makes it simple to use
- Safety features include low water level sensor and over temperature protection
- Stainless steel tank with rounded corners will not trap dirt
- Integral drain simplifies water changes and cleaning
These simple to use heated water baths feature an LED temperature display with an analogue dial for quick and easy set point adjustments. Low water level sensor prevents dry boiling, and over temperature protection system monitors temperature and controls heater in the event of a fault.
Stainless steel tank and sturdy outer casing construction offer excellent durability. Polycarbonate lid provides full view of bath contents and reduces heat loss. Easily drain the tank using the built-in drain port. The heater is mounted beneath the tank, so it is not in the way.
Delivery: Supplied with polycarbonate lid and power cord with plug.
Specifications
- Catalog No:
- STUA12122-71
- STUA12122-81
- STUA12122-91
- Model:
- WB-300-15
- WB-300-24
- WB-300-6
- Electrical:
- 230 VAC, 50 Hz
- 230 VAC, 50 Hz
- 230 VAC, 50 Hz