Order Entry
Ireland
ContactUsLinkComponent
Caprylic/capric triglyceride
Caprylic/capric triglyceride
Supplier:  Spectrum Chemical
Certificates
Caprylic/capric triglyceride
Supplier:  Spectrum Chemical
Certificates
Specification Test Results
  • Saponification Value:
    325 - 360
  • Specific Gravity:
    0.94 - 0.96