Order Entry
Ireland
ContactUsLinkComponent
3-Mercapto-1,2-propanediol ≥97%
Toxic
Irritant
3-Mercapto-1,2-propanediol ≥97%
Catalog # SPCMM1177-25ML
Supplier:  Spectrum Chemical
CAS Number:  
Toxic
Irritant
3-Mercapto-1,2-propanediol ≥97%
Catalog # SPCMM1177-25ML
Supplier:  Spectrum Chemical
CAS Number:  

Specifications

  • Pack type:
    Glass, amber
  • Pk:
    25 mL
  • Seq:
    2315

Specifications

Specification Test Results

  • Assay (Titration By Iodine):
    97% min.
  • Appearance:
    Clear, Colorless

Specification Test Results

About this item

 :
 :108.16 g/mol
Storage Temperature:Refrigerator
UN:2810
ADR:6.1,III
 :GHS06
 :GHS07
CAS Number:96-27-5
Formula:C₃H₈O₂S
MDL Number:MFCD00004879