Order Entry
Ireland
ContactUsLinkComponent
Disodium hydrogen phosphate, dried ≥98.0% USP
Disodium hydrogen phosphate, dried ≥98.0% USP
Catalog # SPCMSO138-2.5KGPL
Supplier:  Spectrum Chemical
CAS Number:  
Disodium hydrogen phosphate, dried ≥98.0% USP
Catalog # SPCMSO138-2.5KGPL
Supplier:  Spectrum Chemical
CAS Number:  

Specifications

  • Pack type:
    Poly bottle
  • Pk:
    2,5 kg
  • Seq:
    4076

Specifications

Specification Test Results

  • Assay (Dried Basis):
    98.0 - 100.5%
  • Loss On Drying:
    max. 5.0 %
  • Insoluble Substances:
    max. 0.4 %
  • Chloride (Cl):
    max. 0.06 %
  • Sulfates (SO₄):
    max. 0.2 %

Specification Test Results

About this item

 :
 :141.96 g/mol
Storage Temperature:Ambient
CAS Number:7558-79-4
Formula:Na₂HPO₄
MDL Number:MFCD00003496