Specifications
- Antibody type:Primary
- Antigen name:MAP2K1IP1
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 750
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Mouse
- Epitope:5-100/124
- Form:Liquid
- Antigen synonyms:MEK-binding partner 1|Mitogen activated protein kinase kinase 1 interacting protein 1|MAP2K1IP1|Mitogen-activated protein kinase kinase 1-interacting protein 1|MP1|MEK partner 1|LAMTOR3|MEK binding partner 1|Late endosomal/lysosomal adaptor and MAPK and MTOR activator 3|LTOR3_HUMAN|Mitogen-activated protein kinase scaffold protein 1|Ragulator complex protein LAMTOR3
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol.
- Molecular weight:13.6 kDa
- Storage temperature:Store at −20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from human MAP2K1IP1
- Tested applications:ICC
- Purification:Purified by Protein A
- Pack type:Vial
- Pk:100 µl
Specifications
About this item
This gene encodes a scaffold protein that functions in the extracellular signal-regulated kinase (ERK) cascade. The protein is localized to late endosomes by the mitogen-activated protein-binding protein-interacting protein, and binds specifically to MAP kinase kinase MAP2K1/MEK1, MAP kinase MAPK3/ERK1, and MAP kinase MAPK1/ERK2. Studies of the orthologous gene in mouse indicate that it regulates late endosomal traffic and cell proliferation. Alternatively spliced transcript variants encoding multiple isoforms have been observed for this gene. A pseudogene of this gene is located on the long arm of chromosome 13. [provided by RefSeq, Aug 2011]
Type: Primary
Antigen: MAP2K1IP1
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 5-100/124
Host: Rabbit
Isotype: IgG
Reactivity: Mouse