Order Entry
Ireland
ContactUsLinkComponent
2'3'-cGAMP ELISA Kit
2'3'-cGAMP ELISA Kit
Catalog # CAYM501700-96
Supplier:  Cayman Chemical
CAS Number:  
undefined
2'3'-cGAMP ELISA Kit
Catalog # CAYM501700-96
Supplier:  Cayman Chemical
CAS Number:  

Specifications

  • Pk:
    96 Tests
  • Assay Type:
    Competitive
  • Format:
    Pre-coated
  • Target protein:
    2'3'-cGAMP
  • Description:
    2'3'-cGAMP ELISA kit
  • Mid point:
    907,7 pg/ml
  • Detection range:
    100 ng/ml - 6,1 pg/ml
  • Detection limit:
    9,6 pg/ml (0,01 pmol/ml)
  • Sensitivity:
    85,3 pg/ml

Specifications

About this item

For the quantification of 2’3’-cGAMP in cell lysates.

  • Measure 2'3'-cGAMP in cell lysates
  • Assay 24 samples in triplicate or 36 samples in duplicate
  • Lower limit of detection (LLOD) is 9.6 pg/ml (0.01 pmol/ml)
  • Run overnight or incubate for just 2 hours without compromising sensitivity
  • Monitor the kinetics of 2'3'-cGAMP formation and hydrolysis in a biological setting
  • Identify compounds that modulate cGAS activity

Cayman’s 2’3’-cGAMP ELISA Kit is a competitive assay that can be used for quantification of 2’3’-cGAMP in cell lysates. This assay has a range from 100 ng/ml to 6,1 pg/ml with a midpoint of approximately 900 pg/ml (50% B/B0) and a sensitivity of approximately 85 pg/ml (80% B/B0). Powered by BIOLOG Life Science Institute.