Anti-HAX1 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
BOSSBS-7626R-A750
: Bioss
- Antibody type:Primary
- Antigen name:HAX1
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 750
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Western blot:Yes
- Epitope:191-279/279
- Form:Liquid
- Gene ID:10456
- Antigen synonyms:HCLS1 associated protein|SCN3|HS1 binding protein 1|Hax1a|HAX 1|HS1 binding protein|HS 1 binding protein|HS1BP1|HS1 associating protein X 1|HCLSBP1|HCLS1 and PKD2 associated protein|HS 1 associated protein X 1|HCLS1 associated protein X 1
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
- Molecular weight:31 kDa
- Storage temperature:Store at ‒20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from human HAX1
- Tested applications:ICC
- Purification:Purified by Protein A
- Pack type:Vial
- Pk:100 µl
RelevanceHAX1 associates with HS1, binding to its N-terminal region. It is also known to associate with PKD2 (involved in polycystic kidney disease) and with cortactin/EMS1. HAX1 is also reported to bind to hairpin structures in vimentin and DNA polymerase beta mRNAs, so may play a role in mRNA stability and transport. It may also function in promoting cell survival. Defects in HAX1 are the cause of autosomal recessive severe congenital neutropenia 3 (SCN3) also called Kostmann disease.
Type: Primary
Antigen: HAX1
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 191-279/279
Host: Rabbit
Isotype: IgG
Reactivity: