Anti-SERCA1 ATPase Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Catalog # BOSSBS-7539R-A750
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:SERCA1 ATPase
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 750
- Flow cytometry:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Rat
- Western blot:Yes
- Epitope:101-200/1001
- Form:Liquid
- Gene ID:487
- Antigen synonyms:Sarcoendoplasmic reticulum calcium ATPase|SR Ca2+-ATPase 1|ATP2A1|Sarcoplasmic reticulum Ca2+-ATPase 1|SERCA1|EC 3.6.3.8|ATP2A|ATPase, Ca2+-transporting fast twitch 1|SR Ca2+ ATPase 1|Calcium transporting ATPase sarcoplasmic reticulum type fast twitch skeletal muscle isoform|Ca-ATPpase|SERCA 1|ATPase Ca++ transporting fast twitch 1|OTTHUMP00000162562|Calcium-transporting ATPase sarcoplasmic reticulum type|ATPase Ca++ transporting cardiac muscle fast twitch 1|fast twitch skeletal muscle isoform|SERCA1 truncated isoform, included|Sarcoplasmic/endoplasmic reticulum calcium ATPase 1|Fast skeletal muscle SR calcium ATPase|OTTHUMP00000162561|Endoplasmic reticulum class 1/2 Ca2+ ATPase|AT2A1_HUMAN|Calcium pump 1
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
- Molecular weight:110 kDa
- Storage temperature:Store at ‒20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from human SERCA1 ATPase
- Tested applications:ICC
- Purification:Purified by Protein A
- Pack type:Vial
- Pk:100 µl
Specifications
About this item
This magnesium-dependent enzyme catalyzes the hydrolysis of ATP coupled with the translocation of calcium from the cytosol to the sarcoplasmic reticulum lumen. Contributes to calcium sequestration involved in muscular excitation/contraction.
Type: Primary
Antigen: SERCA1 ATPase
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 101-200/1001
Host: Rabbit
Isotype: IgG
Reactivity: Rat