Anti-SCYL1BP1 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Catalog # BOSSBS-6643R-A680
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:SCYL1BP1
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 680
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Epitope:201-300/394
- Form:Liquid
- Gene ID:92344
- Antigen synonyms:RAB6-interacting golgin|SCYL1-binding protein 1|GO|GORAB|hNTKL-BP1|NTKL-BP1|SCY1-like 1-binding protein 1|NTKLBP1|NTKL-binding protein 1|SCYL1BP1|SCYL1-BP1|N-terminal kinase-like-binding protein 1
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
- Molecular weight:45 kDa
- Storage temperature:Store at ‒20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from human SCYL1BP1
- Tested applications:ICC
- Purification:Purified by Protein A
- Pack type:Vial
- Pk:100 µl
Specifications
About this item
Defects in GORAB are the cause of geroderma osteodysplasticum (GO) [MIM:231070]; also known as gerodermia osteodysplastica or Walt Disney dwarfism. GO is a rare autosomal recessive disorder characterised by lax, wrinkled skin, joint laxity and a typical face with a prematurely aged appearance. Skeletal signs include severe osteoporosis leading to frequent fractures, malar and mandibular hypoplasia and a variable degree of growth retardation.
Type: Primary
Antigen: SCYL1BP1
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 201-300/394
Host: Rabbit
Isotype: IgG
Reactivity: Human