Specifications
- Antibody type:Primary
- Antigen name:ZNF312B/FEZF1
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 750
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Epitope:199-280/475
- Form:Liquid
- Antigen synonyms:Zinc finger protein 312 like|Fez family zinc finger protein 1|fez like|FEZF1_HUMAN|Zinc finger protein 312B|FEZ family zinc finger 1|fezf1|FEZ
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
- Molecular weight:52 kDa
- Storage temperature:Store at −20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from human ZNF312B
- Tested applications:ICC
- Purification:Purified by Protein A
- Pack type:Vial
- Pk:100 µl
Specifications
About this item
Olfactory sensory neurons contain olfactory receptors, which are G protein-coupled receptor proteins that localize to the cilia and display affinity for and bind to a variety of odor molecules. Olfactory neurons send their axons from the olfactory epithelium to the olfactory bulb, which is covered by the CNS basal lamina. FEZF1 (Fez family zinc finger protein 1), also known as Forebrain Embryonic Zinc Finger and Zinc finger protein 312B, is a 475 amino acid nuclear protein that is expressed in the olfactory epithelium and hypothalamus of mice. In FEZF1-deficient mice, axons of olfactory neurons do not reach the olfactory bulb, suggesting that FEXF1 is required for the penetration of olfactory axons though the basal lamina before innervation of the olfactory bulb. When FEZF1 translocates to the nucleus, it induces KRAS overexpression, resulting in activation of ERK-Signalling. Overexpression of FEZF1 leads to accelerated proliferation in cultured cells and increased tumour mass in mice. There are three isoforms of FEZF1 that are produced as a result of alternative splicing events.
Type: Primary
Antigen: ZNF312B/FEZF1
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 199-280/475
Host: Rabbit
Isotype: IgG
Reactivity: