Anti-TSPAN2 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Catalog # BOSSBS-9411R-A680
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:TSPAN2
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 680
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Form:Liquid
- Gene ID:10100
- Antigen synonyms:Tetraspan TM4SF|Tspan 2|RP4-666F24.2|AW048009|Tetraspanin 2|TSN2_HUMAN|Tetraspanin2|TSN2|Tetraspan 2|B230119D02Rik
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
- Storage temperature:Store at −20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from human TSPAN2
- Purification:Purified by Protein A
- Pack type:Vial
- Pk:100 µl
Specifications
About this item
TSPAN2 is a member of the transmembrane 4 superfamily, also known as the tetraspanin family. TSPAN2 may play a role in Signalling in oligodendrocytes in the early stages of their terminal differentiation into myelin-forming glia and may also function in stabilizing the mature sheath.
Type: Primary
Antigen: TSPAN2
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat