Anti-DRD3 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Catalog # BOSSBS-1743R-A750
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:DRD3
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 750
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Epitope:352-446/446
- Form:Liquid
- Gene ID:13490
- Antigen synonyms:D3R|Dopamine D3 receptor|Drd3|D(3) dopamine receptor
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
- Molecular weight:44 kDa
- Storage temperature:Store at –20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from mouse DRD3
- Tested applications:ICC
- Purification:Purified by Protein A
- Pack type:Vial
- Pk:100 µl
Specifications
About this item
Dopamine receptor whose activity is mediated by G proteins which inhibit adenylyl cyclase. Promotes cell proliferation (By similarity).
Type: Primary
Antigen: DRD3
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 352-446/446
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat