Anti-IL-7Ra/CD127 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
BOSSBS-1540R-A488
: Bioss
- Antibody type:Primary
- Antigen name:IL-7Ra/CD127
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 488
- Flow cytometry:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat
- Western blot:Yes
- Epitope:251-350/459
- Form:Liquid
- Gene ID:3575
- Antigen synonyms:IL-7 receptor subunit alpha|Interleukin-7 receptor subunit alpha|IL-7RA|IL7RA|CD127|ILRA|IL7R|CDW127|IL-7R subunit alpha|IL-7R-alpha
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
- Molecular weight:50 kDa
- Storage temperature:Store at –20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from human IL-7Ra
- Tested applications:ICC
- Purification:Purified by Protein A
- Pack type:Vial
- Pk:100 µl
Receptor for interleukin-7. Also acts as a receptor for thymic stromal lymphopoietin (TSLP).
Type: Primary
Antigen: IL-7Ra/CD127
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 488
Public Immunogen Range: 251-350/459
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat