Anti-GRP75 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Catalog # BOSSBS-1469R-FITC
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:GRP75
- Clonality:Polyclonal
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat
- Western blot:Yes
- Epitope:561-679/679
- Form:Liquid
- Gene ID:3313
- Antigen synonyms:HSPA9|MOT|75 kDa glucose-regulated protein|Mortalin|GRP-75|mt-HSP70|Heat shock 70 kDa protein 9|CRP40|Peptide-binding protein 74|GRP75|MOT2|HSPA9B|CSA|HEL-S-124m|PBP74|MTHSP75|Stress-70 protein, mitochondrial
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
- Molecular weight:70 kDa
- Storage temperature:Store at –20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from human GRP75
- Tested applications:ICC
- Purification:Purified by Protein A
- Pack type:Vial
- Pk:100 µl
Specifications
About this item
Implicated in the control of cell proliferation and cellular aging. May also act as a chaperone.
Type: Primary
Antigen: GRP75
Clonality: Polyclonal
Clone:
Conjugation: FITC
Public Immunogen Range: 561-679/679
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat