Order Entry
Ireland
ContactUsLinkComponent
 
Anti-PSEN1 Mouse Polyclonal Antibody (Alexa Fluor® 680)
Catalog # BOSSBS-0024M-A680
Supplier:  Bioss
Anti-PSEN1 Mouse Polyclonal Antibody (Alexa Fluor® 680)
Catalog # BOSSBS-0024M-A680
Supplier:  Bioss

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Presenilin-1
  • Antigen symbol:
    PSEN1
  • Clonality:
    Polyclonal
  • Conjugation:
    Alexa Fluor® 680
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Mouse
  • Epitope:
    10-80/467
  • Form:
    Liquid
  • Gene ID:
    29192
  • Antigen synonyms:
    Psnl1|Psen1|Presenilin-1|Protein S182|PS-1
  • Modification:
    Unmodified
  • Storage buffer:
    Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
  • Molecular weight:
    34/52 kDa
  • Storage temperature:
    Store at –20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
  • Concentration:
    1 µg/µl
  • Shipping temperature:
    4 °C
  • Immunogen:
    KLH conjugated synthetic peptide derived from rat presenilin 1
  • Tested applications:
    ICC
  • Purification:
    Purified by Protein A
  • Pack type:
    Vial
  • Pk:
    100 µl

Specifications

About this item

Probable catalytic subunit of the gamma-secretase complex, an endoprotease complex that catalyses the intramembrane cleavage of integral membrane proteins such as Notch receptors and APP (beta-amyloid precursor protein). Requires the other members of the gamma-secretase complex to have a protease activity. May play a role in intracellular signaling and gene expression or in linking chromatin to the nuclear membrane. Stimulates cell-cell adhesion though its association with the E-cadherin/catenin complex. Under conditions of apoptosis or calcium influx, cleaves E-cadherin promoting the disassembly of the E-cadherin/catenin complex and increasing the pool of cytoplasmic beta-catenin, thus negatively regulating Wnt signaling. May also play a role in hematopoiesis (By similarity).

Type: Primary
Antigen: Presenilin-1
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 10-80/467
Host: Mouse
Isotype: IgG
Reactivity: Mouse