Anti-Hepatitis C Virus RNA-directed RNA polymerase Rabbit Polyclonal Antibody (Alexa Fluor® 680)
BOSSBS-4858R-A680
: Bioss
- Antibody type:Primary
- Antigen name:Hepatitis C Virus RNA-directed RNA polymerase
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 680
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Virus
- Western blot:Yes
- Epitope:2501-2600/3010
- Form:Liquid
- Antigen synonyms:RNA-directed RNA polymerase|RNA dependent RNA polymerase|p68
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
- Molecular weight:65 kDa
- Storage temperature:Store at −20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from Hepatitis C Virus RNA-directed RNA polymerase
- Tested applications:ICC
- Purification:Purified by Protein A
- Pack type:Vial
- Pk:100 µl
The RNA directed RNA polymerase is also known as non-structural protein NS5B. NS5B is a 65 kDa protein that resembles other viral RNA polymerases. Hepatitis C virus (HCV) replication is thought to occur in membrane bound replication complexes. These complexes transcribe the positive strand and the resulting minus strand is used as a template for the synthesis of genomic RNA. There are two viral proteins involved in the reaction, NS3 and NS5B.
Type: Primary
Antigen: Hepatitis C Virus RNA-directed RNA polymerase
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 2501-2600/3010
Host: Rabbit
Isotype: IgG
Reactivity: Virus