Anti-IL-2 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Catalog # BOSSBS-4586R-A680
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:IL-2
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 680
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Western blot:Yes
- Epitope:81-169/169
- Form:Liquid
- Gene ID:16183
- Antigen synonyms:Il2|Interleukin-2|TCGF|Il-2|T-cell growth factor
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
- Molecular weight:15 kDa
- Storage temperature:Store at −20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from mouse IL-2
- Tested applications:ICC
- Purification:Purified by Protein A
- Pack type:Vial
- Pk:100 µl
Specifications
About this item
Produced by T-cells in response to antigenic or mitogenic stimulation, this protein is required for T-cell proliferation and other activities crucial to regulation of the immune response. Can stimulate B-cells, monocytes, lymphokine-activated killer cells, natural killer cells, and glioma cells.
Type: Primary
Antigen: IL-2
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 81-169/169
Host: Rabbit
Isotype: IgG
Reactivity: