Anti-TPX2 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Catalog # BOSSBS-4285R-A750
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:TPX2
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 750
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Mouse
- Western blot:Yes
- Epitope:551-650/747
- Form:Liquid
- Gene ID:22974
- Antigen synonyms:TPX2_HUMAN|HCTP 4|Targeting protein for XKLP2|DIL-2|C20ORF1|repp86|REPP 86|HCA 519|HCTP4|Dferentially expressed in cancerous and non-cancerous lung cells 2|Protein FLS353|TPX2 microtubule associated protein homolog|TPX 2|DIL2|Preferentially expressed in colorectal cancer|Hepatocellular carcinoma-associated antigen 519|C20orf2|p100|Restricted expression proliferation associated protein 100|TPX2 microtubule associated homolog|Dferentially expressed in cancerous and noncancerous lung cells 2|Dferentially expressed in lung cells|GD:C20orf1|Dferentially expressed in lung cells 2|Restricted expression proliferation-associated protein 100|FLS353|Chromosome 20 Open Reading Frame 1|TPX2|DIL 2|Hepatocellular carcinoma associated antigen 519|HCA519
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
- Molecular weight:86 kDa
- Storage temperature:Store at −20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from human TPX2
- Tested applications:ICC
- Purification:Purified by Protein A
- Pack type:Vial
- Pk:100 µl
Specifications
About this item
TPX2 is essential for spindle pole and mitotic spindle formulation, playing a role in the S and G2 phases of mitosis. It is also critical for the function of protein kinase Aurora A. TPX2 autophosphorylates Aurora A, targeting it to the spindle apparatus and allowing it to function in eukaryotic meiotic and mitotic cell cycles. TPX2 has also been found to be highly expressed in cancer cells.
Type: Primary
Antigen: TPX2
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 551-650/747
Host: Rabbit
Isotype: IgG
Reactivity: Mouse