Anti-DEGS1 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
BOSSBS-4057R-A680
: Bioss
- Antibody type:Primary
- Antigen name:DEGS1
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 680
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Epitope:21-120/323
- Form:Liquid
- Gene ID:653121
- Antigen synonyms:DEGS 1|Membrane fatty acid lipid desaturase|Sphingolipid delta4 desaturase DES1.DEGS1_HUMAN|Des 1|Degenerative spermatocyte homolog lipid desaturase|Degenerative spermatocyte homolog 1|Dihydroceramide desaturase|Degenerative spermatocyte homolog 1 lipid desaturase|MIG15v Migration inducing gene 15 protein|MLD|MGC5079|Sphingolipid delta 4 desaturase|FADS7|Membrane lipid desaturase|DES1|DEGS
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
- Molecular weight:35 kDa
- Storage temperature:Store at −20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from human DEGS1
- Tested applications:ICC
- Purification:Purified by Protein A
- Pack type:Vial
- Pk:100 µl
DEGS1 is a member of the membrane fatty acid desaturase family which is responsible for inserting double bonds into specific positions in fatty acids. It contains three His containing consensus motifs that are characteristic of a group of membrane fatty acid desaturases. It has sphingolipid-delta-4-desaturase activity and converts D-erythro-sphinganine to D-erythro-sphingosine (E-sphing-4-enine).
Type: Primary
Antigen: DEGS1
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 21-120/323
Host: Rabbit
Isotype: IgG
Reactivity: