Order Entry
Ireland
ContactUsLinkComponent
Anti-Vimentin Ser82 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Anti-Vimentin Ser82 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Catalog # BOSSBS-3472R-A680
Supplier:  Bioss
Anti-Vimentin Ser82 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Catalog # BOSSBS-3472R-A680
Supplier:  Bioss

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Vimentin Ser82
  • Clonality:
    Polyclonal
  • Conjugation:
    Alexa Fluor® 680
  • Flow cytometry:
    Yes
  • Host:
    Rabbit
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Sheep,
    Human,
    Mouse
  • Western blot:
    Yes
  • Form:
    Liquid
  • Gene ID:
    7431
  • Antigen synonyms:
    VIMphospho S82|OTTHUMP00000019224|VIME_HUMAN|Vimentin|VIM|FLJ36605
  • Modification:
    Phosphorylation (Ser82)
  • Storage buffer:
    Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
  • Molecular weight:
    51 kDa
  • Storage temperature:
    Store at −20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
  • Concentration:
    1 µg/µl
  • Shipping temperature:
    4 °C
  • Immunogen:
    KLH conjugated synthetic phosphopeptide derived from human Vimentin around the phosphorylation site of Ser82
  • Tested applications:
    ICC
  • Purification:
    Purified by Protein A
  • Pack type:
    Vial
  • Pk:
    100 µl

Specifications

About this item

This gene encodes a member of the intermediate filament family. Intermediate filamentents, along with microtubules and actin microfilaments, make up the cytoskeleton. The protein encoded by this gene is responsible for maintaining cell shape, integrity of the cytoplasm, and stabilising cytoskeletal interactions. It is also involved in the immune response, and controls the transport of low-density lipoprotein (LDL)-derived cholesterol from a lysosome to the site of esterification. It functions as an organiser of a number of critical proteins involved in attachment, migration, and cell signaling. Mutations in this gene causes a dominant, pulverulent cataract.

Type: Primary
Antigen: Vimentin Ser82
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range:
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse; Sheep