Order Entry
Ireland
ContactUsLinkComponent
 
Anti-SHC Tyr427 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Catalog # BOSSBS-3412R-A680
Supplier:  Bioss
Anti-SHC Tyr427 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Catalog # BOSSBS-3412R-A680
Supplier:  Bioss

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    SHC Tyr427
  • Clonality:
    Polyclonal
  • Conjugation:
    Alexa Fluor® 680
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Mouse
  • Western blot:
    Yes
  • Form:
    Liquid
  • Gene ID:
    6464
  • Antigen synonyms:
    SHC|SH2 domain protein C1|SHC1|Src homology 2 domain-containing-transforming protein C1|SHC-transforming protein A|SHC-transforming protein 1|SHC-transforming protein 3|SHCA
  • Modification:
    Phosphorylation (Tyr427)
  • Storage buffer:
    Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
  • Molecular weight:
    63 kDa
  • Storage temperature:
    Store at −20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
  • Concentration:
    1 µg/µl
  • Shipping temperature:
    4 °C
  • Immunogen:
    KLH conjugated synthetic phosphopeptide derived from human SHC around the phosphorylation site of Tyr427
  • Tested applications:
    ICC
  • Purification:
    Purified by Protein A
  • Pack type:
    Vial
  • Pk:
    100 µl

Specifications

About this item

Signaling adapter that couples activated growth factor receptors to signaling pathways. Participates in a signaling cascade initiated by activated KIT and KITLG/SCF. Isoform p46Shc and isoform p52Shc, once phosphorylated, couple activated receptor tyrosine kinases to Ras via the recruitment of the GRB2/SOS complex and are implicated in the cytoplasmic propagation of mitogenic signals. Isoform p46Shc and isoform p52Shc may thus function as initiators of the Ras signaling cascade in various non-neuronal systems. Isoform p66Shc does not mediate Ras activation, but is involved in signal transduction pathways that regulate the cellular response to oxidative stress and life span. Isoform p66Shc acts as a downstream target of the tumor suppressor p53 and is indispensable for the ability of stress-activated p53 to induce elevation of intracellular oxidants, cytochrome c release and apoptosis. The expression of isoform p66Shc has been correlated with life span (By similarity). Participates in signaling downstream of the angiopoietin receptor TEK/TIE2, and plays a role in the regulation of endothelial cell migration and sprouting angiogenesis.

Type: Primary
Antigen: SHC Tyr427
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range:
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse