v6.5 Mouse Embryonic Stem Cells Cell Lysate (Embryonic Normal)
Catalog # NOVUNBP1-97433
Supplier: Novus Biologicals
CAS Number:
Specifications
- Cell Format:Cryopreserved
- Pk:0,25 mg
- Tested applications:WB, ImmunoPrecipitation
Specifications
About this item
v6.5 Mouse embryonic stem cells Cell Lysate (Embryonic Normal).
Tested Applications: Western Blot, Immunoprecipitation.