Anti-IBP160 Rabbit Polyclonal Antibody (Cy5)
Catalog # BOSSBS-15530R-CY5
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:IBP160
- Clonality:Polyclonal
- Conjugation:Cy5®
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Western blot:Yes
- Epitope:1-100/1485
- Form:Liquid
- Antigen synonyms:AQR|IBP160|Intron-binding protein aquarius|functional spliceosome-associated protein 164|aquarius homolog (mouse)|AQR_HUMAN|DKFZp686B23123|fSAP164|aquarius|Intron-binding protein of 160 kDa|KIAA0560
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
- Molecular weight:171 kDa
- Storage temperature:Store at –20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from human IBP160
- Tested applications:ICC
- Purification:Purified by Protein A
- Pack type:Vial
- Pk:100 µl
Specifications
About this item
IBP160.
Type: Primary
Antigen: IBP160
Clonality: Polyclonal
Clone:
Conjugation: Cy5
Public Immunogen Range: 1-100/1485
Host: Rabbit
Isotype: IgG
Reactivity: