Anti-MAPT Chicken Polyclonal Antibody
Catalog # ROCK200-901-X29
Supplier: Rockland Immunochemicals
Specifications
- Antibody type:Primary
- Antigen name:Microtubule-associated protein tau
- Antigen symbol:MAPT
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Chicken
- ImmunoChemistry:Yes
- Reactivity:Human,Rat
- Western blot:Yes
- Format:IgY
- Form:Liquid (sterile filtered)
- Gene ID:4137
- Antigen synonyms:Pired helical filament-tau|Neurofibrillary tangle protein|PHF-tau
- Storage buffer:0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2
- Storage temperature:-20°C
- Shipping temperature:Dry Ice
- Immunogen:Anti-Tau Antibody was produced in chickens by repeated immunizations with a recombinant full length human tau expressed in and purified from E.Coli.
- Pk:100 µl