Order Entry
Ireland
ContactUsLinkComponent
 
Newborn Bovine Serum, MP Biomedicals
  ICNA092912154
undefined
Newborn Bovine Serum, MP Biomedicals
  ICNA092912154

 

  • Sterility:
    Sterile
  • Description:
    Newborn Bovine Serum, 500 ml
  • Endotoxin:
    30 EU/ml
  • Storage temperature:
    Store at –20 °C
  • Application:
    Cell culture and organ preservation applications
  • Concentration:
    Protein: 4,4 - 7,0 g/dl
  • Pk:
    500 mL

 

 

This serum is produced from blood collected from newly born calves between 1 to 14 days of age.

  • Cell culture and organ preservation
  • Culture Media
  • Suitable for cell culture
  • Shelf Life 36 months

Newborn Bovine Serum (NBS) is useful in a wide range of cell culture and organ preservation applications. For example, it is used in cornea preservation and cryopreservation of bovine embryos. NBS is used in cell culture to support the maturation of oocytes, to study the function of tubular epithelial cells, and to develop primary cultures of cells such as visceral adipocytes.