Human IL-13 PicoKine; ELISA Kit
Catalog # BSBTEK0424
Supplier: Boster Bio
CAS Number:
Specifications
- Pk:1 KIT
- Assay Type:Sandwich
- Host:
- Primary antibody reactivity:Human
- Target protein:IL-13
- Description:Human IL-13 PicoKine; ELISA kit, 96 wells/kit
- Environmentally Preferable:
Specifications
About this item
Sandwich High Sensitivity ELISA kit for Quantitative Detection of Human IL-13.
- Sensitivity, Detect single digit picogram level of target protein
- Specificity, Thoroughly tested for cross-reactivity with family members
- Stability, Guaranteed to remain stable for up to 12 months with proper storage
- Selection, 1000+ kits available, covering most common ELISA targets
- Service, On-demand technical support directly from ELISA experts
Detect Human IL13 with <10 pg/ml sensitivity. Format: 96-well plate with removable strips. Compatible samples: cell culture supernates, cell lysates, serum and plasma (heparin, EDTA). This is a TMB colorimetric sandwich ELISA kit with short assay time and fast experiment set up. IL13 tissue specificity:
96wells/kit, with removable strips.