Order Entry
Ireland
ContactUsLinkComponent
Primary Human Melanocyte Cells
Primary Human Melanocyte Cells
Catalog # CELSCSC2HM1
Supplier:  CELL SYSTEMS
CAS Number:  
Primary Human Melanocyte Cells
Catalog # CELSCSC2HM1
Supplier:  CELL SYSTEMS
CAS Number:  

Specifications

  • Cell Format:
    Cryopreserved
  • Pk:
    1 KIT
  • Single Donor/Pooled:
    Pooled
  • Description:
    Human melanocyte cells
  • Storage conditions:
    Upon receipt, store in liquid nitrogen
  • Storage buffer:
    Frozen in Cell Systems cell freezing medium (4Z0705)

Specifications

About this item

Our primary human cells offer unmatched consistency and a more biologically relevant tool than other cell types. Our cells are antibody-free and come from a deep inventory to help researchers with reproducibility and improved human insights.

  • Highly proliferative and healthy (~85% viability)
  • Antibody free and more biologically relevant
  • Offered with high cell count (~1 million cells per vial)
  • Well-cited in scientific journals
  • Ideal for 2D and 3D cultures, microfluidic devices and co-cultures

Primary Human Melanocyte Cells (CSC2HM1) were isolated from foreskin tissue of multiple neonatal donors. This cell is antibody free. Each vial will initiate a 25 to 75 cm² cell culture. These primary cells are isolated, frozen and qualified using Cell Systems Certified Media and Reagents--which are specified for use in the research environment. Cell Systems primary cells are exclusively from normal, site-specific human tissue specimen, and available in frozen vial or proliferating flask.

These cells were originated using Cell Systems Complete Serum-Free Medium (SF-4Z0-500), and subsequently grown and passaged in Cell Systems Complete Medium (4Z0-500). They are available at Passage 3 (<12 cumulative population doubling)] cryopreserved in Cell Systems Cell Freezing Medium™ (4Z0-705). This vial will initiate a Passage 4 cell culture in a 75 cm² flask. Each vial contains more than 1×106 cells.

Each vial of cells is shipped with vials of Bac-Off® (antibiotic) and CultureBoost™ (animal derived growth factors) or CultureBoost-R™ (human recombinant growth factors) at no additional cost.

Standard Tests:
HIV Test (donor level HIV AB EIA) -Negative
HIV PCR Test (frozen cell pool by CLIA Licensed Clinical Lab) - Negative
Mycoplasma spp. (frozen cells) (ATCC method by CLIA Licensed Clinical Lab) - Negative.

Delivery: Shipped on dry ice.
Caution: For research use only.