- Cell Format:Cryopreserved
- Pk:1 KIT
- Single Donor/Pooled:Single donor
- Description:Human coronary artery smooth muscle cells
- Storage conditions:Upon receipt, store in liquid nitrogen
- Storage buffer:Frozen in Cell Systems cell freezing medium (4Z0705)
Our primary human cells offer unmatched consistency and a more biologically relevant tool than other cell types. Our cells are antibody-free and come from a deep inventory to help researchers with reproducibility and improved human insights.
- Highly proliferative and healthy (~85% viability)
- Antibody free and more biologically relevant
- Offered with high cell count (~1 million cells per vial)
- Well-cited in scientific journals
- Ideal for 2D and 3D cultures, microfluidic devices and co-cultures
Primary Human Coronary Artery Smooth Muscle Cells (ACBRI443) were initiated from normal human coronary arterial tissue. This cell is antibody free. Each vial will initiate a 25 to 75 cm² cell culture. These primary cells are isolated, frozen and qualified using Cell Systems Certified Media and Reagents--which are specified for use in the research environment. Cell Systems primary cells are exclusively from normal, site-specific human tissue specimen, and available in frozen vial or proliferating flask.
These cells were originated using Cell Systems Complete Serum-Free Medium (SF-4Z0-500), and subsequently grown and passaged in Cell Systems Complete Medium (4Z0-500). They are available at Passage 3 (<12 cumulative population doublings) cryopreserved in Cell Systems Cell Freezing Medium™ (4Z0-705). This vial will initiate a Passage 4 cell culture in a 75 cm² flask. Each vial contains more than 1×106 cells.
Each vial of cells is shipped with vials of Bac-Off® (antibiotic) and CultureBoost™ (animal derived growth factors) or CultureBoost-R™ (human recombinant growth factors) at no additional cost.
Standard Tests:
HIV Serologic Test (donor level HIV AB EIA) - Negative
HIV PCR Test (frozen cell pool by CLIA Licensed Clinical Lab) - Negative
Mycoplasma spp. (frozen cells) (ATCC method by CLIA Licensed Clinical Lab) - Negative.
Characterisation:
Inducible expression of CD 62E (E-Selectin) >90% positive by immunofluorescence
Cytoplasmic VWF/Factor VIII >95% positive by immunofluorescence
Cytoplasmic uptake of Di-L-Ac-LDL >95% positive by immunofluorescence.