PIG® Medical Absorbent Mat Pads, New Pig
Catalog # NEWPMATE1111
Supplier: NEW PIG
Specifications
- Description:Medical absorbent mat pads
- L×W:500×900 mm
- Seq:01
- Absorbency:57,6 L
- Pk:80
Specifications
About this item
Prevent slips and falls in the operating theatre with super-absorbent medical mat.
- Keeps floors dry — poly-backing keeps floors safe and dry during wet operating procedures
- Ideal for hospital floors — contain and absorb spills from procedures like knee and hip operations where saline fluids are used in abundance for irrigation
- Super absorbent — made of a non-wetting top layer to keep the standing surface dry, a super-absorbent central layer and a non-slip backing that grips to the floor
- Reduce slips and falls — place with white, floor-gripping side down to add traction and reduce slip hazards
- Minimise tripping hazards — no more tripping over bunched-up blankets; increases focus on the patient and not the work environment
- Reduces clean-up time — can be easily lifted and disposed of after use
- Pre-cut pads are simple to use – place mat on clean, dry floor with white, floor-gripping side down to catch liquids as they accumulate; when mat is saturated, lift from floor and remove for disposal in accordance with local regulations
Material:
- Top layer: Three-dimensional perforated LDPE
- Core: Air-laid cellulose with polypropylene
- Base layer: Polyethylene high-grip film