2-Bromo-5-iodo-3-methylbenzoic acid ≥95%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1022983-52-3
:
Formula:C₈H₆BrIO₂
:340.9405 g/mol
Frequently Bought Together
Benzyl acetate for synthesis, Sigma-Aldrich®
Supplier: Merck
Bis(2-butoxyethyl)ether for synthesis, Sigma-Aldrich®
Supplier: Merck
Methanol 99.8%, ECD tested for pesticide analysis for residue analysis
Supplier: Thermo Fisher Scientific
Chloroform
Supplier: Roth Carl
Isoamyl alcohol 98%, pure
Supplier: Thermo Fisher Scientific
Ethanol denatured ≥96%, meets analytical specification of Ph.Eur. (denaturated with 1%iPr-OH,1% MEK,0.001% Bitrex®)
Supplier: Thermo Fisher Scientific
Toluene (Reag. Ph. Eur.) for analysis, ACS, ISO 1 * 2,5 L
Supplier: PanReac AppliChem
Pyridine ≥99%
Supplier: Thermo Fisher Scientific
Benzonitrile 99+% for spectroscopy
Supplier: Thermo Fisher Scientific
Ethyl acetate ≥99.5% ACS
Supplier: Thermo Fisher Scientific
Polyvinyl alcohol, M.W. ∼ 124,000 99-100%, hydrolysed
Supplier: Thermo Fisher Scientific
tert-Butyl methyl ether for analysis
Supplier: Thermo Fisher Scientific
Dichloromethane 99.6% stabilised ACS
Supplier: Thermo Fisher Scientific
Tetrahydrofuran ≥99.9% inhibitor-free, CHROMASOLV™ Plus for HPLC, Riedel-de Haën™
Supplier: Honeywell Chemicals
2-Propanol 99.6% ACS, meets analytical specifications of Reagent Ph. Eur.
Supplier: Thermo Fisher Scientific