


Hydrogen peroxide 35%, INTEROX® SG-35-Plus
Supplier: SOLVAY
Certificates
About this item
CAS Number:7722-84-1
:
Formula:H₂O₂
MDL Number:MFCD00011333
Boiling Pt:107,4 °C
Density:0,8835
Storage Temperature:Ambient
EINECS:231-765-0
UN:2014
ADR:5.1 (8),II
:34.01 g/mol
:GHS03
:GHS05
:GHS07
Specification Test Results
- Assay (GC):35,0-35,7%
- stability (1h,96°):≤ 3g/kg*h
- non-volatile matter (105°):<2ppm
- calcium:≤0,05 ppm
- iron:≤0,01 ppm
- aluminium:≤0,05 ppm
- sodium:≤0,5 ppm
- chloride:≤0,05 ppm
- phosphate (PO43-):≤1 ppm
- sulfate:≤0,5 ppm
- cadmium:≤0,002 ppm
- chromium:≤0,002 ppm
- cobalt:≤0,002 ppm
- copper:≤0,002 ppm
- manganese:≤0,002 ppm
- magnesium:≤0,01 ppm
- nickel:≤0,002 ppm
- tin:≤0,002 ppm
- zinc:≤0,002 ppm
- silicon:≤0,01 ppm
- nitrate:≤3 ppm
Specification Test Results
Related Information
Certificates
Search by Lot Numberspec_INTEROX(R) SG-35-PLUS Hydrogen Peroxide Biocidal Product Grade EMEA Solvey
Product specification
Frequently Bought Together