UV to white light converter plates
UVLTUV/WL26
: UVITEC
- Description:UV/white light converter screen 260×210 mm
- Environmentally Preferable:
- Pk:1
The specially designed converter plate converts UV radiation from the transilluminator using a white light diffuser to enable Coomassie® Blue or silver stained protein gels to be viewed.
- Handles are situated on opposite sides of the converter plate for easy handling